Iminodibenzyl, 97%, Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
CAS: 494-19-9 | C14H13N | 195.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA122070250
Price (MYR)
320.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
320.00
Add to cart
Chemical Identifiers
CAS12045-63-5
IUPAC Namediboron titanium
Molecular FormulaB2Ti
InChI KeyQYEXBYZXHDUPRC-UHFFFAOYSA-N
SMILES[B].[B].[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentOxygen 0.5% max