Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
CAS: 68-94-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA122010050
Price (MYR)
325.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
325.00
Add to cart
Chemical Identifiers
CAS7789-09-5
IUPAC Namediammonium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2H8N2O7
InChI KeyJOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
Appearance (Form)Crystalline powder
Loss on drying=<3.0 %
Chloride (Cl)=<0.005 %
Calcium (Ca)=<0.002 %