Allopurinol, 98%
Allopurinol, 98%
Allopurinol, 98%
Thermo Scientific Chemicals
Allopurinol, 98%
CAS: 315-30-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA121830250
Price (MYR)
494.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
494.00
Add to cart
Chemical Identifiers
CAS142-62-1
IUPAC Namehexanoic acid
Molecular FormulaC6H12O2
InChI KeyFUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILESCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4150 to 1.4180 (20°C, 589 nm)