Allopurinol, 98%
Allopurinol, 98%
Allopurinol, 98%
Thermo Scientific Chemicals
Allopurinol, 98%
CAS: 315-30-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA121830050
Price (MYR)
250.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
250.00
Add to cart
Chemical Identifiers
CAS7774-34-7
IUPAC Namecalcium hexahydrate dichloride
Molecular FormulaCaCl2H12O6
InChI KeyQHFQAJHNDKBRBO-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Alkalinity=<0.03 % (as Ca(OH)2)
Sulfate (SO4)=<0.05 %
Titration Complexometric97.0 to 103.0 %
Insoluble matter=<0.025 % (in water and ppt by NH4OH)