4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
CAS: 121-34-6 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA121350250
Price (MYR)
359.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
359.00
Add to cart
Chemical Identifiers
CAS13446-03-2
IUPAC Namemanganese(2+) dibromide
Molecular FormulaBr2Mn
InChI KeyRJYMRRJVDRJMJW-UHFFFAOYSA-L
SMILES[Mn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Impurity contentH₂O <0.3%