Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA120860010
Price (MYR)
1,260.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,260.00
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Concentration1.00-1.05 (OD)
Polydispersity Index≤0.2
Buffer0.1 mM phosphate buffered saline (0.01X PBS)
lambda-max (UV-Vis)518-528 nm