1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
CAS: 629-11-8 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA120650010
Price (MYR)
462.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
462.00
Add to cart
Chemical Identifiers
CAS77-71-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
Loss on drying=<0.5 % (60°C, 2 h, 1 mmHg)
Appearance (Form)Crystalline powder or crystals
Melting point175°C to 180°C