Guaiacol, 99+%, Thermo Scientific Chemicals
Guaiacol, 99+%, Thermo Scientific Chemicals
Guaiacol, 99+%, Thermo Scientific Chemicals
Guaiacol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Guaiacol, 99+%, Thermo Scientific Chemicals
CAS: 90-05-1 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number FSA120190010
Price (MYR)
671.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
671.00
Add to cart
Chemical Identifiers
CAS72-57-1
IUPAC Nametetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC34H24N6Na4O14S4
InChI KeyWMYVHJWZUUEZNE-ARWFNKCKSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to brown-black
Dye content>=60 %
UV spectrumComplies
Appearance (Form)Powder