Glycidol, 96%, Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
CAS: 556-52-5 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA120051000
Price (MYR)
493.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
493.00
Add to cart
Specifications
Chemical Name or MaterialGlycidol
CAS556-52-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing genetic defects.
May cause cancer.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
View more