2-Furoic acid, 98%, Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA119821000
Price (MYR)
371.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
371.00
Add to cart
Chemical Identifiers
CAS5356-71-8
IUPAC Name1,3-diphenyl-1H-pyrazol-5-amine
Molecular FormulaC15H13N3
InChI KeySXOFMEWDEKEVJU-UHFFFAOYSA-N
SMILESNC1=CC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)125.5-134.5?C
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder