Furfurylamine, 99+%, Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA119802500
Price (MYR)
475.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
475.00
Add to cart
Chemical Identifiers
CAS432-25-7
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥90.0%
Refractive Index1.4850-1.5050 @ 20?C
Free acid (titration)≤3.0%