Fumaric acid, 99+%, Thermo Scientific Chemicals
Fumaric acid, 99+%, Thermo Scientific Chemicals
Fumaric acid, 99+%, Thermo Scientific Chemicals
Fumaric acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fumaric acid, 99+%, Thermo Scientific Chemicals
CAS: 110-17-8 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number FSA119751000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS150-13-0
IUPAC Name4-aminobenzoic acid
Molecular FormulaC7H7NO2
InChI KeyALYNCZNDIQEVRV-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥98.5
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Appearance (Color)White to pale brown