Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
CAS: 3473-63-0 | CH4N2·C2H4O2 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA119715000
Price (MYR)
1,125.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,125.00
Add to cart
Chemical Identifiers
CAS5580-80-3
IUPAC Name2,3,4,5-tetrafluoroaniline
Molecular FormulaC6H3F4N
InChI KeyBEECAQIHCYTZHC-UHFFFAOYSA-N
SMILESNC1=CC(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%