Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA119160050
Price (MYR)
562.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
562.00
Add to cart
Chemical Identifiers
CAS35231-44-8
IUPAC Name4-(bromomethyl)-7-methoxy-2H-chromen-2-one
Molecular FormulaC11H9BrO3
InChI KeyCTENSLORRMFPDH-UHFFFAOYSA-N
SMILESCOC1=CC=C2C(CBr)=CC(=O)OC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
HPLC>=96.0 %
Melting point212°C to 217°C