Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA118450010
Price (MYR)
923.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
923.00
Add to cart
Chemical Identifiers
CAS10034-93-2
IUPAC Namehydrazine; sulfuric acid
Molecular FormulaH6N2O4S
InChI KeyZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILESNN.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<50 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Crystalline powder
Titration Iodimetric>=99.0 %
Heavy metals (as Pb)=<20 ppm