Ethylenediaminetetraacetic acid, 99%, pure
Ethylenediaminetetraacetic acid, 99%, pure
Ethylenediaminetetraacetic acid, 99%, pure
Ethylenediaminetetraacetic acid, 99%, pure
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, 99%, pure
CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Catalog number FSA118432500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS80126-53-0
IUPAC Name(2S)-2-amino-3-(2-methylphenyl)propanoic acid
Molecular FormulaC10H13NO2
InChI KeyNHBKDLSKDKUGSB-VIFPVBQESA-N
SMILESCC1=CC=CC=C1C[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Optical Rotation+2? ± 0.5? (c=1 in 5M HCl)