Ethylcyclohexane, 99+%, Thermo Scientific Chemicals
Ethylcyclohexane, 99+%, Thermo Scientific Chemicals
Ethylcyclohexane, 99+%, Thermo Scientific Chemicals
Ethylcyclohexane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylcyclohexane, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA118311000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylcyclohexane
Name Note99+%
CAS1678-91-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.