Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
CAS: 140-88-5 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA117980250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56-92-8
IUPAC Namedihydrogen 2-(1H-imidazol-5-yl)ethan-1-amine dichloride
Molecular FormulaC5H11Cl2N3
InChI KeyPPZMYIBUHIPZOS-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C)
Sulfated ash=<0.1 %
Appearance (Color)White
Appearance (Form)Crystalline powder