Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
CAS: 140-88-5 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA117980025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS28489-45-4
IUPAC Name6-methyl-5-nitro-1,2-dihydropyridin-2-one
Molecular FormulaC6H6N2O3
InChI KeyAJDDHNXZBGZBJN-UHFFFAOYSA-N
SMILESCC1=C(C=CC(=O)N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Melting Point (clear melt)233.0-240.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)Pale yellow to pale brown