Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
CAS: 531-75-9 | C15H16O9 | 340.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
50 g
Packaging:
Glass bottle
Catalog number FSA117830100
Price (MYR)
424.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
424.00
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.9995% (metals basis)
Appearance (Color)Silvery white
FormSlug, 3.175mm (0.125in) diameter.