Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
CAS: 531-75-9 | C15H16O9 | 340.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA117830050
Price (MYR)
305.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
305.00
Add to cart
Chemical Identifiers
CAS320-97-8
IUPAC Name4-fluorobenzene-1,2-dicarboxylic acid
Molecular FormulaC8H5FO4
InChI KeyOMCXTFVBNCFZMY-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(F)C=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%