Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
CAS: 106-89-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 mL
1 L
Packaging:
Glass bottle
Catalog number FSA117780025
Price (MYR)
2,266.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,266.00
Add to cart
Chemical Identifiers
CAS4780-79-4
IUPAC Name(naphthalen-1-yl)methanol
Molecular FormulaC11H10O
InChI KeyPBLNHHSDYFYZNC-UHFFFAOYSA-N
SMILESOCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystalline powder or powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)57-64?C