Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
CAS: 106-89-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA117780010
Price (MYR)
779.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
779.00
Add to cart
Chemical Identifiers
CAS36589-58-9
IUPAC Namedisodium propane-1,3-disulfonate
Molecular FormulaC3H6Na2O6S2
InChI KeyDKGJFKPIUSHDIT-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)CCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated Ash56.4-58.1%
FormPowder
Loss on Drying<5.0% (200?C/constant wt.)
Appearance (Color)White
Assay (ex Sulphated Ash)>98.5 to <101.5% (dry wt. basis)