Ellagic acid, 97%
Ellagic acid, 97%
Ellagic acid, 97%
Thermo Scientific Chemicals
Ellagic acid, 97%
CAS: 476-66-4 | C14H6O8 | 302.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
50 g
Packaging:
Glass bottle
Catalog number FSA117740100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS823-85-8
IUPAC Namehydrogen (4-fluorophenyl)hydrazine chloride
Molecular FormulaC6H8ClFN2
InChI KeyFEKUXLUOKFSMRO-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Argentometric>=96.0 %
Appearance (Color)White to brown