Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number FSA117550250
Price (MYR)
1,457.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,457.00
Add to cart
Chemical Identifiers
CAS78-94-4
IUPAC Namebut-3-en-2-one
Molecular FormulaC4H6O
InChI KeyFUSUHKVFWTUUBE-UHFFFAOYSA-N
SMILESCC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=93 %
Water=<5 %
Stabilizer=<0.7 % Acetonitrile
Stabilizer0.2 to 0.5 % Hydroquinone