Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number FSA117550050
Price (MYR)
452.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
452.00
Add to cart
Chemical Identifiers
CAS109-79-5
IUPAC Namebutane-1-thiol
Molecular FormulaC4H10S
InChI KeyWQAQPCDUOCURKW-UHFFFAOYSA-N
SMILESCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=99.0 %
Appearance (Form)Liquid