5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
CAS: 69-78-3 | C14H8N2O8S2 | 396.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA117540050
Price (MYR)
519.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
519.00
Add to cart
Specifications
Chemical Name or Material5, 5'-Dithiobis-(2-nitrobenzoic Acid)
CAS69-78-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.