9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA117200010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS100-19-6
IUPAC Name1-(4-nitrophenyl)ethan-1-one
Molecular FormulaC8H7NO3
InChI KeyYQYGPGKTNQNXMH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Appearance (Color)Yellow
Identification (FTIR)Conforms
Melting Point (clear melt)76.0-83.0?C