1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA117110010
Price (MYR)
502.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Specifications
Chemical Name or Material1, 4-Dioxane
Melting Point12.0°C
Name Note99+%
CAS123-91-1
CAS Min %99.0