Dimethyl Phthalate, 99%, Thermo Scientific Chemicals
Dimethyl Phthalate, 99%, Thermo Scientific Chemicals
Dimethyl Phthalate, 99%, Thermo Scientific Chemicals
Dimethyl Phthalate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl Phthalate, 99%, Thermo Scientific Chemicals
CAS: 131-11-3 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA116642500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl phthalate
CAS131-11-3
Density1.1900g/mL
EINECS Number205-011-6
Boiling Point282°C