2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 576-26-1 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA116572500
Price (MYR)
306.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
306.00
Add to cart
Chemical Identifiers
CAS31138-65-5
IUPAC Namesodium (3R,4S,5R,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
Molecular FormulaC7H13NaO8
InChI KeyFMYOMWCQJXWGEN-BMZZJELJSA-M
SMILES[Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min.
Chloride content0.01% max.
Assay (unspecified)Sulfate: 0.1% max.
Water Content (Karl Fischer Titration)14.0% max.