Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA116420050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-91-2
IUPAC Name(oxiran-2-yl)methyl 2-methylprop-2-enoate
Molecular FormulaC7H10O3
InChI KeyVOZRXNHHFUQHIL-UHFFFAOYNA-N
SMILESCC(=C)C(=O)OCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<30 APHA