N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 6638-79-5 | C2H8ClNO | 97.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA116340050
Price (MYR)
298.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
298.00
Add to cart
Chemical Identifiers
CAS5460-29-7
IUPAC Name2-(3-bromopropyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC11H10BrNO2
InChI KeyVKJCJJYNVIYVQR-UHFFFAOYSA-N
SMILESBrCCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystalline powder or powder
Assay (GC)≥97.5%
CommentMay contain a small amount of dark particulates caused by trace bromine in the crystals
Identification (FTIR)Conforms
View more
Coupling reagent
RUO – Research Use Only