3-Dimethylaminopropylchloride hydrochloride, 99%, Thermo Scientific Chemicals
3-Dimethylaminopropylchloride hydrochloride, 99%, Thermo Scientific Chemicals
3-Dimethylaminopropylchloride hydrochloride, 99%, Thermo Scientific Chemicals
3-Dimethylaminopropylchloride hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Dimethylaminopropylchloride hydrochloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA115911000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS872041-86-6
IUPAC Name(5-fluoropyridin-3-yl)boronic acid
Molecular FormulaC5H5BFNO2
InChI KeyFVEDGBRHTGXPOK-UHFFFAOYSA-N
SMILESOB(O)C1=CC(F)=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)White to off-white