4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
Thermo Scientific Chemicals
4-Dimethylaminocinnamaldehyde, 98%
CAS: 6203-18-5 | C11H13NO | 175.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA115790050
Price (MYR)
508.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
508.00
Add to cart
Chemical Identifiers
CAS10531-80-3
IUPAC Name6-methyl-1,3-benzoxazole
Molecular FormulaC8H7NO
InChI KeySZWNDAUMBWLYOQ-UHFFFAOYSA-N
SMILESCC1=CC=C2N=COC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
GC>=99.50 %
Melting point29°C to 31°C
Appearance (Color)Amber