4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA115760250
Price (MYR)
336.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
336.00
Add to cart
Chemical Identifiers
CAS13777-25-8
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min