N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 mL
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number FSA115690250
Price (MYR)
3,327.00
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
3,327.00
Add to cart
Chemical Identifiers
CAS3637-63-6
IUPAC Namecyclooctylmethanol
Molecular FormulaC9H18O
InChI KeyZHPBLHYKDKSZCQ-UHFFFAOYSA-N
SMILESOCC1CCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4820-1.4870 @ 20?C