N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA115690050
Price (MYR)
307.00
Add to cart
Quantity:
5 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
307.00
Add to cart
Chemical Identifiers
CAS25697-55-6
IUPAC Name[3-(trifluoromethyl)phenyl]methanethiol
Molecular FormulaC8H7F3S
InChI KeyCQIQWIMXCPTQPJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(CS)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>88.0%
Appearance (Color)Clear, colorless
Refractive Index1.4860-1.4940 @ 20?C