N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA115690025
Price (MYR)
793.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
793.00
Add to cart
Chemical Identifiers
CAS635-12-1
IUPAC Name1,4-dihydroanthracene-1,4-dione
Molecular FormulaC14H8O2
InChI KeyLSOTZYUVGZKSHR-UHFFFAOYSA-N
SMILESO=C1C=CC(=O)C2=C1C=C1C=CC=CC1=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
FormPowder
Assay (HPLC)≥92.0%