N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA115690010
Price (MYR)
411.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
411.00
Add to cart
Chemical Identifiers
CAS4733-39-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Infrared spectrumConforms
Appearance (Form)Powder
Melting point277°C to 285°C (Dec)
Titration with HClO497.5 to 102.5 %