2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals
CAS: 91-10-1 | C8H10O3 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA115591000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS99-02-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
Refractive Index1.5475-1.5515 @ 20?C