3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals
3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals
3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals
3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals
CAS: 530-59-6 | C11H12O5 | 224.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA115550050
Price (MYR)
561.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
561.00
Add to cart
Chemical Identifiers
CAS409-21-2
IUPAC Namemethanidylidynesilylium
Molecular FormulaCSi
InChI KeyHBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES[C-]#[Si+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5% SiC
Impurity contentFe₂O₃: ≤0.5%
Impurity contentFree C: ≤0.5%
Particle SizeConforms to FEPA Micro grit standard
Particle Sizemean (d50) 9.3 ? 1 ?m