3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
CAS: 93-07-2 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA115451000
Price (MYR)
444.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Chemical Identifiers
CAS4392-24-9
IUPAC Name[(1E)-3-bromoprop-1-en-1-yl]benzene
Molecular FormulaC9H9Br
InChI KeyRUROFEVDCUGKHD-QPJJXVBHSA-N
SMILESBrC\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Appearance (Color)White to pale cream to cream to yellow to orange to dark brown to brown
Assay (GC)≥94.0%
Refractive Index1.6160-1.6220 @ 20?C