3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
CAS: 93-07-2 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA115450250
Price (MYR)
259.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
259.00
Add to cart
Chemical Identifiers
CAS99-89-8
IUPAC Name4-(propan-2-yl)phenol
Molecular FormulaC9H12O
InChI KeyYQUQWHNMBPIWGK-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink to brown
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder or crystals or fused solid
Melting point58°C to 63°C