2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number FSA115311000
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS16853-85-3
IUPAC Namelithium(1+) alumanuide
Molecular FormulaAlH4Li
InChI KeyOCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES[Li+].[AlH4-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Appearance (Form)Powder
Assay>=94 % (Gasvolumetric)