2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA115310250
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1120-06-5
IUPAC Namedecan-2-ol
Molecular FormulaC10H22O
InChI KeyACUZDYFTRHEKOS-UHFFFAOYNA-N
SMILESCCCCCCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %