Diethyl malonate, 99+%, Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
CAS: 105-53-3 | C7H12O4 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA114450250
Price (MYR)
231.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
231.00
Add to cart
Chemical Identifiers
CAS89-98-5
IUPAC Name2-chlorobenzaldehyde
Molecular FormulaC7H5ClO
InChI KeyFPYUJUBAXZAQNL-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5650 to 1.5670 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow