Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA114290250
Price (MYR)
2,403.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,403.00
Add to cart
Chemical Identifiers
CAS104-95-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)low melting crystalline mass
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals and/or chunks or
View more
For analysis
RUO – Research Use Only