Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
CAS: 1115-30-6 | C10H16O5 | 216.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA113950250
Price (MYR)
407.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
407.00
Add to cart
Chemical Identifiers
CAS146-68-9
IUPAC Name3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular FormulaC19H13ClIN5O2
InChI KeyJORABGDXCIBAFL-UHFFFAOYSA-M
SMILES[Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %