N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
100 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number FSA113900250
Price (MYR)
32,933.00
Add to cart
Quantity:
25 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
32,933.00
Add to cart
Chemical Identifiers
CAS59649-56-8
IUPAC Name2,3-diaminophenol
Molecular FormulaC6H8N2O
InChI KeyPCAXITAPTVOLGL-UHFFFAOYSA-N
SMILESNC1=C(N)C(O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Melting point161°C to 165°C
Appearance (Color)Light brown to dark brown
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Coupling reagent
RUO – Research Use Only